Gene Transcripts
- Gene Symbol
- AUP1
- Entrez Gene ID
- 550
- Gene Name
- ancient ubiquitous protein 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 74526648 - 74529897 on Build GRCh38
- UniGene ID
- Hs.411480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181575.4 |
NP_853553.1
|
6, 7 |
947 |
|
NR_126510.1 |
|
6, 7 |
947 |
|
NR_126511.1 |
|
5, 6 |
1143 |
|
XM_005264392.3 |
XP_005264449.1
|
5, 6 |
892 |
|
GenBank mRNA |
AF100746.1 |
- |
6, 7 |
720 |
|
AF100753.1 |
- |
6, 7 |
747 |
|
AF100754.1 |
- |
5, 6 |
945 |
|
AF165515.1 |
- |
6, 7 |
727 |
|
AK023983.1 |
- |
6, 7 |
725 |
|
AK293121.1 |
- |
6, 7 |
766 |
|
AK304886.1 |
- |
5, 6 |
889 |
|
AK308231.1 |
- |
5, 6 |
946 |
|
AK310159.1 |
- |
5, 6 |
945 |
|
BC001658.1 |
- |
6, 7 |
810 |
3050204 |
BC033646.2 |
- |
6, 7 |
762 |
|
CR749661.1 |
- |
5, 6 |
818 |
|
View Full Product Details