Gene Transcripts

Gene Symbol
Olr459
Entrez Gene ID
295729
Gene Name
olfactory receptor 459
Gene Aliases
Chromosome Location
Chr. 3: 73177656 - 73178585 on Build Rnor_6.0
UniGene ID
Rn.142975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000293.1 NP_001000293.1 1 616

View Full Product Details