Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321207.1 | NP_001308136.1 | 5, 6 | 767 | |
NM_015355.3 | NP_056170.2 | 6, 7 | 836 | ||
XM_006721794.3 | XP_006721857.1 | 4, 5 | 278 | ||
GenBank mRNA | AK074333.1 | - | 1, 2 | 73 | |
AK290014.1 | - | 6, 7 | 807 | ||
BC015704.1 | - | 6, 7 | 678 | 3908024 | |
BC018583.1 | - | 5, 6 | 710 | ||
D63881.2 | - | 6, 7 | 793 |