Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015954.3 | NP_057038.2 | 6 | 772 | |
GenBank mRNA | AF132960.1 | - | 6 | 633 | |
AK024614.1 | - | 6 | 588 | ||
AK222541.1 | - | 6 | 588 | ||
AK296697.1 | - | 6 | 529 | ||
AK309182.1 | - | 6 | 688 | ||
BC041328.1 | - | 6 | 772 | ||
BC056234.1 | - | 6 | 614 |