Gene Transcripts
- Gene Symbol
- YIPF1
- Entrez Gene ID
- 54432
- Gene Name
- Yip1 domain family member 1
- Gene Aliases
- DJ167A19.1, FinGER1
- Chromosome Location
- Chr. 1: 53851719 - 53889831 on Build GRCh38
- UniGene ID
- Hs.11923
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018982.4 |
NP_061855.1
|
5, 6 |
655 |
|
NR_036639.1 |
|
5, 6 |
655 |
|
NR_036640.1 |
|
4, 5 |
435 |
|
NR_135075.1 |
|
3, 4 |
372 |
|
XM_017001499.1 |
XP_016856988.1
|
4, 5 |
413 |
|
XM_017001500.1 |
XP_016856989.1
|
4, 5 |
476 |
|
XM_017001501.1 |
XP_016856990.1
|
3, 4 |
257 |
|
GenBank mRNA |
AK000823.1 |
- |
3, 4 |
320 |
|
AK300240.1 |
- |
4, 5 |
517 |
|
AK315183.1 |
- |
5, 6 |
633 |
|
AL713668.1 |
- |
4, 5 |
366 |
|
BC064590.1 |
- |
5, 6 |
621 |
5749036 |
BI757212.1 |
- |
5, 6 |
652 |
|
BX405963.2 |
- |
5, 6 |
647 |
|
CN308348.1 |
- |
5, 6 |
656 |
|
View Full Product Details