Gene Transcripts
- Gene Symbol
- Olr1365
- Entrez Gene ID
- 287081
- Gene Name
- olfactory receptor 1365
- Gene Aliases
- Chromosome Location
- Chr. 10: 12213881 - 12214822 on Build Rnor_6.0
- UniGene ID
- Rn.142201
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_214824.1 |
NP_999989.1
|
|
596 |
|
- Gene Symbol
- LOC100912159
- Entrez Gene ID
- 100912159
- Gene Name
- olfactory receptor 1361-like
- Gene Aliases
- Chromosome Location
- Chr. 10: 12428753 - 12434386 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details