Gene Transcripts

Gene Symbol
Henmt1
Entrez Gene ID
100363095
Gene Name
HEN1 methyltransferase homolog 1 (Arabidopsis)
Gene Aliases
RGD1306230
Chromosome Location
Chr. 2: 211740571 - 211753718 on Build Rnor_6.0
UniGene ID
Rn.47322
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037655.1 NP_001032744.2 254
GenBank mRNA BC107926.1 - 254

View Full Product Details