Gene Transcripts
- Gene Symbol
- HDHD2
- Entrez Gene ID
- 84064
- Gene Name
- haloacid dehalogenase like hydrolase domain containing 2
- Gene Aliases
- 3110052N05Rik, HEL-S-301
- Chromosome Location
- Chr. 18: 47107402 - 47150520 on Build GRCh38
- UniGene ID
- Hs.465041
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032124.4 |
NP_115500.1
|
2, 3 |
242 |
|
XM_005258371.4 |
XP_005258428.1
|
2, 3 |
262 |
|
XM_011526227.2 |
XP_011524529.1
|
3, 4 |
318 |
|
XM_011526228.2 |
XP_011524530.1
|
3, 4 |
316 |
|
XM_017026039.1 |
XP_016881528.1
|
3, 4 |
355 |
|
GenBank mRNA |
AK292098.1 |
- |
2, 3 |
242 |
|
AL136681.1 |
- |
2, 3 |
233 |
|
BC038100.2 |
- |
2, 3 |
242 |
5266439 |
CR533487.1 |
- |
1, 2 |
109 |
|
FJ224322.1 |
- |
2, 3 |
233 |
|
View Full Product Details