Gene Transcripts

Gene Symbol
Vom1r101
Entrez Gene ID
286892
Gene Name
vomeronasal 1 receptor 101
Gene Aliases
V1rb13, V1rb7, VN7, Vnr7
Chromosome Location
Chr. 4: 119398198 - 119399130 on Build Rnor_6.0
UniGene ID
Rn.10227
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173112.2 NP_775135.2 1 222
GenBank mRNA AY510287.1 - 1 222
U36786.1 - 1 140

View Full Product Details