Gene Transcripts
- Gene Symbol
- A730020M07Rik
- Entrez Gene ID
- 100503044
- Gene Name
- RIKEN cDNA A730020M07 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 121634936 - 121646453 on Build GRCm38
- UniGene ID
- Mm.65320
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042746.1 |
- |
4 |
1406 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
NM_176992.1 |
- |
|
1405 |
|
Chr. - - [-] |
View Full Product Details