Gene Transcripts

Gene Symbol
Gm13051
Entrez Gene ID
626316
Gene Name
predicted gene 13051
Gene Aliases
OTTMUSG00000010104, OTTMUSG00000010173
Chromosome Location
Chr. 4: 146097312 - 146126624 on Build GRCm38
UniGene ID
Mm.270762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037926.2 NP_001033015.1 1 72
GenBank mRNA BC108336.1 - 1 50
CD547545.1 - 1 72

View Full Product Details