Gene Transcripts

Gene Symbol
Gmeb2
Entrez Gene ID
83635
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
Chromosome Location
Chr. 3: 176756006 - 176791960 on Build Rnor_6.0
UniGene ID
Rn.40949
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031803.1 NP_113991.1 2 207
GenBank mRNA AF059273.1 - 2 207
AF205778.1 - 98
AF205779.1 - 98
AF205780.1 - 98
FQ211504.1 - 237

View Full Product Details