Gene Transcripts

Gene Symbol
Isoc1
Entrez Gene ID
66307
Gene Name
isochorismatase domain containing 1
Gene Aliases
2610034N03Rik
Chromosome Location
Chr. 18: 58659482 - 58679570 on Build GRCm38
UniGene ID
Mm.182574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025478.3 NP_079754.2 5 2034
GenBank mRNA AY035213.1 - 5 2034

View Full Product Details