Gene Transcripts

Gene Symbol
ALDOB
Entrez Gene ID
229
Gene Name
aldolase, fructose-bisphosphate B
Gene Aliases
ALDB, ALDO2
Chromosome Location
Chr. 9: 101420560 - 101435780 on Build GRCh38
UniGene ID
Hs.530274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000035.3 NP_000026.2 3 234
GenBank mRNA AK290795.1 - 3 228
BC029399.1 - 3 228 4593670
X01098.1 - 3 218
X02747.1 - 3 277

View Full Product Details