Gene Transcripts

Gene Symbol
Olfr1496
Entrez Gene ID
258991
Gene Name
olfactory receptor 1496
Gene Aliases
MOR127-1
Chromosome Location
Chr. 19: 13780548 - 13781614 on Build GRCm38
UniGene ID
Mm.377780
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146989.2 NP_667200.2 1 140
GenBank mRNA BC137878.1 - 1 140

View Full Product Details