Gene Transcripts

Gene Symbol
Olfr447
Entrez Gene ID
258990
Gene Name
olfactory receptor 447
Gene Aliases
MOR261-1
Chromosome Location
Chr. 6: 42911525 - 42912457 on Build GRCm38
UniGene ID
Mm.377779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146988.1 NP_667199.1 1 893
GenBank mRNA BC104271.1 - 1 893
BC104272.1 - 1 891

View Full Product Details