Gene Transcripts
- Gene Symbol
- Cntnap5b
- Entrez Gene ID
- 241175
- Gene Name
- contactin associated protein-like 5B
- Gene Aliases
- C230078M14Rik, Caspr5-2
- Chromosome Location
- Chr. 1: 99772247 - 100486066 on Build GRCm38
- UniGene ID
- Mm.251301
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172851.2 |
NP_766439.2
|
24 |
5027 |
|
GenBank mRNA |
AK082656.1 |
- |
17 |
3915 |
|
View Full Product Details