gRNA Design Details

Target DNA Sequence
TTAGTACTTCTACAGACTCC
PAM Sequence
AGG
Target locus
Chr.11: 46808090 - 46808112 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CKAP5
Entrez Gene ID
9793
Gene Name
cytoskeleton associated protein 5
Gene Aliases
CHTOG, MSPS, TOG, TOGp, ch-TOG
Chromosome Location
Chr.11: 46743534 - 46846308 on GRCh38
UniGene ID
Hs.201253
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001008938.3 NP_001008938.1 8
NM_014756.3 NP_055571.2 8