gRNA Design Details
- Target DNA Sequence
- TTAGTACTTCTACAGACTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 46808090 - 46808112 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CKAP5
- Entrez Gene ID
-
9793
- Gene Name
- cytoskeleton associated protein 5
- Gene Aliases
-
CHTOG, MSPS, TOG, TOGp, ch-TOG
- Chromosome Location
-
Chr.11: 46743534 - 46846308 on GRCh38
- UniGene ID
- Hs.201253
- Species
-
Human