gRNA Design Details
- Target DNA Sequence
- CAGTCTTCTTGAAGCGCATG
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 60564407 - 60564429 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
81.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Rbm22
- Entrez Gene ID
-
66810
- Gene Name
- RNA binding motif protein 22
- Gene Aliases
-
8430430L24Rik
- Chromosome Location
-
Chr.18: 60560786 - 60572729 on GRCm38
- UniGene ID
- Mm.275106
- Species
-
Mouse