gRNA Design Details
- Target DNA Sequence
- GTTCTGCCTGTTGTAGGTGT
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 150700944 - 150700966 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
67.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RBM22
- Entrez Gene ID
-
55696
- Gene Name
- RNA binding motif protein 22
- Gene Aliases
-
Cwc2, ZC3H16, fSAP47
- Chromosome Location
-
Chr.5: 150690790 - 150701107 on GRCh38
- UniGene ID
- Hs.713564
- Species
-
Human