gRNA Design Details
- Target DNA Sequence
- CGCCATGGATACTGTACTTT
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 58205283 - 58205305 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fbxo32
- Entrez Gene ID
-
67731
- Gene Name
- F-box protein 32
- Gene Aliases
-
4833442G10Rik, AI430017, ATROGIN1, Gm20361, MAFbx
- Chromosome Location
-
Chr.15: 58175879 - 58214892 on GRCm38
- UniGene ID
- Mm.25683
- Species
-
Mouse