gRNA Design Details
- Target DNA Sequence
- AATAGTGCGTTAACAGGCTC
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 48981002 - 48981024 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Dzip3
- Entrez Gene ID
-
224170
- Gene Name
- DAZ interacting protein 3, zinc finger
- Gene Aliases
-
2310047C04Rik, 2A-HUB, 6430549P11Rik, A230104G20
- Chromosome Location
-
Chr.16: 48924228 - 48994153 on GRCm38
- UniGene ID
- Mm.275138
- Species
-
Mouse