gRNA Design Details
- Target DNA Sequence
- ACGATCAGTCGAATCTGACT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 54466109 - 54466131 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.15/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fnip1
- Entrez Gene ID
-
216742
- Gene Name
- folliculin interacting protein 1
- Gene Aliases
-
A730024A03Rik, AI838773, AW557298
- Chromosome Location
-
Chr.11: 54438146 - 54518241 on GRCm38
- UniGene ID
- Mm.34087
- Species
-
Mouse