gRNA Design Details

Target DNA Sequence
CTATTTCGGGGACGCAATTC
PAM Sequence
AGG
Target locus
Chr.14: 22929283 - 22929305 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 77.21/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRMT5
Entrez Gene ID
10419
Gene Name
protein arginine methyltransferase 5
Gene Aliases
HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs
Chromosome Location
Chr.14: 22920511 - 22929585 on GRCh38
UniGene ID
Hs.367854
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001039619.2 NP_001034708.1 1
NM_001282953.1 NP_001269882.1 1
NM_001282954.1 NP_001269883.1 1
NM_001282955.1 NP_001269884.1 1
NM_001282956.1 NP_001269885.1 1
NM_006109.4 NP_006100.2 1
XM_017020927.1 XP_016876416.1 1