gRNA Design Details
- Target DNA Sequence
- CCAGTGTGTCAGCTATTTCG
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 54517266 - 54517288 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prmt5
- Entrez Gene ID
-
27374
- Gene Name
- protein arginine N-methyltransferase 5
- Gene Aliases
-
Jbp1, Skb1
- Chromosome Location
-
Chr.14: 54507182 - 54517470 on GRCm38
- UniGene ID
- Mm.196585
- Species
-
Mouse