gRNA Design Details
- Target DNA Sequence
- GAACCTGCTAAGAATCGGCC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 54516535 - 54516557 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prmt5
- Entrez Gene ID
-
27374
- Gene Name
- protein arginine N-methyltransferase 5
- Gene Aliases
-
Jbp1, Skb1
- Chromosome Location
-
Chr.14: 54507182 - 54517470 on GRCm38
- UniGene ID
- Mm.196585
- Species
-
Mouse