gRNA Design Details

Target DNA Sequence
TGCCAATAAGGACAACGAAG
PAM Sequence
TGG
Target locus
Chr.5: 76877721 - 76877743 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 92.19/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
S100Z
Entrez Gene ID
170591
Gene Name
S100 calcium binding protein Z
Gene Aliases
Gm625, S100-zeta
Chromosome Location
Chr.5: 76849953 - 76952882 on GRCh38
UniGene ID
Hs.482563
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_130772.3 NP_570128.2 4
XM_011543239.2 XP_011541541.1 4
XM_011543240.2 XP_011541542.1 4
XM_011543241.2 XP_011541543.1 4
XM_011543244.2 XP_011541546.1 4
XM_011543245.2 XP_011541547.1 3
XM_017009171.1 XP_016864660.1 4
XM_017009172.1 XP_016864661.1 4
XM_017009173.1 XP_016864662.1 3