gRNA Design Details
- Target DNA Sequence
- AGTCCTCACTCTCCTCCAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 52269020 - 52269042 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
77.76/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Foxo1
- Entrez Gene ID
-
56458
- Gene Name
- forkhead box O1
- Gene Aliases
-
AI876417, Afxh, FKHR, Fkhr1, Foxo1a
- Chromosome Location
-
Chr.3: 52268337 - 52350109 on GRCm38
- UniGene ID
- Mm.29891
- Species
-
Mouse