gRNA Design Details
- Target DNA Sequence
- AAGAGTCGGCAAGTCTGTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 123513281 - 123513303 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FBXO32
- Entrez Gene ID
-
114907
- Gene Name
- F-box protein 32
- Gene Aliases
-
Fbx32, MAFbx
- Chromosome Location
-
Chr.8: 123497887 - 123541253 on GRCh38
- UniGene ID
- Hs.403933
- Species
-
Human