gRNA Design Details

Target DNA Sequence
CTAGCTCTAAGTGAACCACG
PAM Sequence
AGG
Target locus
Chr.2: 48454706 - 48454728 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 97.27/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PPP1R21
Entrez Gene ID
129285
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
CCDC128, KLRAQ1
Chromosome Location
Chr.2: 48440769 - 48515392 on GRCh38
UniGene ID
Hs.654619
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001135629.2 NP_001129101.1 3
NM_001193475.1 NP_001180404.1 3
NM_152994.4 NP_694539.1 3
NR_024188.2 3
XM_017003301.1 XP_016858790.1 3
XM_017003302.1 XP_016858791.1 3