gRNA Design Details
- Target DNA Sequence
- AATCAAGAACTATCCGAACA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 223959718 - 223959740 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MRPL44
- Entrez Gene ID
-
65080
- Gene Name
- mitochondrial ribosomal protein L44
- Gene Aliases
-
COXPD16, L44MT, MRP-L44
- Chromosome Location
-
Chr.2: 223943224 - 223967714 on GRCh38
- UniGene ID
- Hs.203559
- Species
-
Human