gRNA Design Details
- Target DNA Sequence
- GATGCATGTCGGCTTCTCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 219246756 - 219246778 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK16
- Entrez Gene ID
-
8576
- Gene Name
- serine/threonine kinase 16
- Gene Aliases
-
KRCT, MPSK, PKL12, TSF1
- Chromosome Location
-
Chr.2: 219245469 - 219250337 on GRCh38
- UniGene ID
- Hs.153003
- Species
-
Human