gRNA Design Details
- Target DNA Sequence
- CTAGCACTAAGTGAACCTCG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 88545432 - 88545454 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ppp1r21
- Entrez Gene ID
-
73825
- Gene Name
- protein phosphatase 1, regulatory subunit 21
- Gene Aliases
-
1110018J12Rik, AI426045, AW550781, Ccdc128, Klraq1
- Chromosome Location
-
Chr.17: 88530124 - 88588367 on GRCm38
- UniGene ID
- Mm.271817
- Species
-
Mouse