gRNA Design Details
- Target DNA Sequence
- CGGAGTTGAAAGCAGATCAA
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 31852822 - 31852844 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- COPRS
- Entrez Gene ID
-
55352
- Gene Name
- coordinator of PRMT5 and differentiation stimulator
- Gene Aliases
-
C17orf79, COPR5, HSA272196, TTP1
- Chromosome Location
-
Chr.17: 31851865 - 31859307 on GRCh38
- UniGene ID
- Hs.462729
- Species
-
Human