gRNA Design Details

Target DNA Sequence
AAGAAACGTAAAGTCAAAGC
PAM Sequence
AGG
Target locus
Chr.3: 108616583 - 108616605 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 83.93/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
DZIP3
Entrez Gene ID
9666
Gene Name
DAZ interacting zinc finger protein 3
Gene Aliases
PPP1R66, UURF2, hRUL138
Chromosome Location
Chr.3: 108589490 - 108694846 on GRCh38
UniGene ID
Hs.409210
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_014648.3 NP_055463.1 5
XM_005247914.3 XP_005247971.1 5
XM_005247915.3 XP_005247972.1 5
XM_005247916.3 XP_005247973.1 6
XM_005247917.3 XP_005247974.1 5
XM_005247918.3 XP_005247975.1 5
XM_005247919.3 XP_005247976.1 5
XM_017007540.1 XP_016863029.1 5