Gene Transcripts

Gene Symbol
WNT8B
Entrez Gene ID
7479
Gene Name
Wnt family member 8B
Gene Aliases
Chromosome Location
Chr. 10: 100463055 - 100483642 on Build GRCh38
UniGene ID
Hs.421281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003393.3 NP_003384.2 3 263
GenBank mRNA AB073637.1 - 3 261
CN276452.1 - 3 263
X91940.1 - 2 83
Y11094.1 - 3 286

View Full Product Details