Gene Transcripts

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
Chromosome Location
Chr. 7: 146116035 - 148420998 on Build GRCh38
UniGene ID
Hs.655684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014141.5 NP_054860.1 8 1803
XM_017011950.1 XP_016867439.1 8 2129
GenBank mRNA AB020675.1 - 8 1312
AF319045.1 - 8 1427
AK315113.1 - 8 1392
BC093780.1 - 8 1330 7939625
BC113373.1 - 8 1330 8322425
KJ534811.1 - 7 1176

View Full Product Details