Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022371.3 | NP_071766.2 | 4 | 843 | |
GenBank mRNA | AJ299403.1 | - | 4 | 732 | |
AJ299441.1 | - | 4 | 732 | ||
AK025998.1 | - | 4 | 722 | ||
AK130132.1 | - | 2 | 162 | ||
AK299966.1 | - | 3 | 440 | ||
AK316530.1 | - | 3 | 452 | ||
BC001085.1 | - | 4 | 741 | 2959819 | |
BC007571.1 | - | 4 | 732 | 3029304 | |
BC011746.1 | - | 4 | 728 | 3343691 | |
BC018292.1 | - | 4 | 733 | 3457259 | |
BC088368.1 | - | 3 | 470 | ||
CR457354.1 | - | 4 | 691 |