Gene Transcripts

Gene Symbol
BANF2
Entrez Gene ID
140836
Gene Name
barrier to autointegration factor 2
Gene Aliases
BAF-L, BAF2, BAFL, C20orf179
Chromosome Location
Chr. 20: 17693675 - 17735872 on Build GRCh38
UniGene ID
Hs.115366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014977.3 NP_001014977.2 3 293
NM_001159495.1 NP_001152967.1 3 217
NM_178477.4 NP_848572.3 4 456
XM_005260668.3 XP_005260725.1 5 829
XM_011529170.1 XP_011527472.1 4 666
XM_011529171.2 XP_011527473.1 4 468
GenBank mRNA AA394107.1 - 3 256
AW022487.1 - 3 225
BC054871.1 - 3 297 6592641
BC062735.1 - 4 447 6591888

View Full Product Details