Gene Transcripts

Gene Symbol
CTAGE1
Entrez Gene ID
64693
Gene Name
cutaneous T-cell lymphoma-associated antigen 1
Gene Aliases
CT21.1, CT21.2, CTAGE, CTAGE-1, CTAGE-2
Chromosome Location
Chr. 18: 22413601 - 22417915 on Build GRCh38
UniGene ID
Hs.406709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172241.2 NP_758441.2 1 1546
GenBank mRNA AF273058.1 - 1 1554
BC031065.1 - 1 1548
BC132871.1 - 1 1479 40146893

View Full Product Details