Gene Transcripts

Gene Symbol
HLA-DRB6
Entrez Gene ID
3128
Gene Name
major histocompatibility complex, class II, DR beta 6 (pseudogene)
Gene Aliases
Chromosome Location
Chr. 6: 32552713 - 32560002 on Build GRCh38
UniGene ID
Hs.723344
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001298.1 2 270
GenBank mRNA AF005487.1 - 2 270
L76565.1 - 2 270
U33547.1 - 2 290

View Full Product Details