Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68226515-68227015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2

View Full Product Details