Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68231102-68231598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2 XM_011531609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908452 c.515G>A p.G172E Chr.4: 68231374-68231374 on Build GRCh38
3940920 c.516G>A p.G172G Chr.4: 68231373-68231373 on Build GRCh38
4734692 c.520C>T p.Q174* Chr.4: 68231369-68231369 on Build GRCh38
1229788 c.526G>T p.A176S Chr.4: 68231363-68231363 on Build GRCh38
5866629 c.577G>A p.E193K Chr.4: 68231312-68231312 on Build GRCh38
View More

View Full Product Details