Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68235677-68236191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2 XM_011531609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605055 c.240G>A p.K80K Chr.4: 68236151-68236151 on Build GRCh38
4734694 c.213A>C p.T71T Chr.4: 68236178-68236178 on Build GRCh38
3605054 c.240+1G>A p.? Chr.4: 68236150-68236150 on Build GRCh38
6357981 c.212C>G p.T71R Chr.4: 68236179-68236179 on Build GRCh38
3605053 c.257A>G p.Q86R Chr.4: 68236053-68236053 on Build GRCh38
View More

View Full Product Details