Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68236010-68236513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2 XM_011531609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125632 c.182A>C p.N61T Chr.4: 68236209-68236209 on Build GRCh38
4734694 c.213A>C p.T71T Chr.4: 68236178-68236178 on Build GRCh38
6512285 c.201A>T p.S67S Chr.4: 68236190-68236190 on Build GRCh38
4524929 c.129G>A p.K43K Chr.4: 68236262-68236262 on Build GRCh38
380184 c.197C>A p.A66D Chr.4: 68236194-68236194 on Build GRCh38
View More

View Full Product Details