Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105146533-105147025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
UniGene
Hs.367639
Gene Chromosome Location
Chr.4: 105145875-105279803 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1

View Full Product Details