Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19379405-19379905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6
Entrez Gene ID
6194
Gene Name
ribosomal protein S6
Gene Aliases
S6
UniGene
Hs.408073
Gene Chromosome Location
Chr.9: 19376256-19380237 on Build GRCh38
Transcripts
NM_001010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864628 c.71T>A p.L24H Chr.9: 19379554-19379554 on Build GRCh38
3906486 c.97G>A p.A33T Chr.9: 19379528-19379528 on Build GRCh38
5506279 c.159G>A p.L53L Chr.9: 19379466-19379466 on Build GRCh38
5031141 c.36C>G p.C12W Chr.9: 19379589-19379589 on Build GRCh38
6182928 c.23C>T p.P8L Chr.9: 19379602-19379602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140144551 Chr.9: 19379736-19379736 on Build GRCh38
rs774463196 Chr.9: 19379482-19379482 on Build GRCh38
rs758793587 Chr.9: 19379510-19379510 on Build GRCh38
rs530117224 Chr.9: 19379437-19379437 on Build GRCh38
rs769973213 Chr.9: 19379466-19379466 on Build GRCh38
rs1002866157 Chr.9: 19379880-19379880 on Build GRCh38
rs770388104 Chr.9: 19379596-19379596 on Build GRCh38
rs928533939 Chr.9: 19379707-19379707 on Build GRCh38
rs200489092 Chr.9: 19379632-19379632 on Build GRCh38
rs940960824 Chr.9: 19379691-19379691 on Build GRCh38
rs940909247 Chr.9: 19379815-19379816 on Build GRCh38
rs140661033 Chr.9: 19379743-19379743 on Build GRCh38
rs769490290 Chr.9: 19379641-19379641 on Build GRCh38
rs762812062 Chr.9: 19379628-19379628 on Build GRCh38
rs768596769 Chr.9: 19379551-19379551 on Build GRCh38
rs370530821 Chr.9: 19379542-19379542 on Build GRCh38
rs760228833 Chr.9: 19379506-19379506 on Build GRCh38
rs759233340 Chr.9: 19379604-19379604 on Build GRCh38
rs182404061 Chr.9: 19379695-19379695 on Build GRCh38
rs553185340 Chr.9: 19379658-19379658 on Build GRCh38
rs751897427 Chr.9: 19379513-19379513 on Build GRCh38
rs530252736 Chr.9: 19379788-19379788 on Build GRCh38
rs11541616 Chr.9: 19379607-19379607 on Build GRCh38
rs773188115 Chr.9: 19379586-19379586 on Build GRCh38
rs1032182095 Chr.9: 19379742-19379742 on Build GRCh38
rs922906621 Chr.9: 19379854-19379854 on Build GRCh38
rs913274064 Chr.9: 19379626-19379626 on Build GRCh38
rs1029578768 Chr.9: 19379815-19379815 on Build GRCh38
rs563637618 Chr.9: 19379812-19379812 on Build GRCh38
rs902831131 Chr.9: 19379533-19379533 on Build GRCh38
rs764836018 Chr.9: 19379605-19379605 on Build GRCh38
rs570889419 Chr.9: 19379839-19379839 on Build GRCh38
rs202025440 Chr.9: 19379637-19379637 on Build GRCh38
rs771972723 Chr.9: 19379581-19379581 on Build GRCh38
rs752590926 Chr.9: 19379471-19379472 on Build GRCh38
rs901538186 Chr.9: 19379711-19379711 on Build GRCh38
rs149265455 Chr.9: 19379601-19379601 on Build GRCh38
rs371732122 Chr.9: 19379828-19379828 on Build GRCh38
rs200536268 Chr.9: 19379481-19379481 on Build GRCh38
rs531584522 Chr.9: 19379568-19379568 on Build GRCh38
rs761317554 Chr.9: 19379660-19379660 on Build GRCh38
rs369659987 Chr.9: 19379667-19379667 on Build GRCh38
rs762219638 Chr.9: 19379609-19379609 on Build GRCh38
rs141607232 Chr.9: 19379493-19379493 on Build GRCh38
rs902883440 Chr.9: 19379773-19379773 on Build GRCh38
rs368144672 Chr.9: 19379753-19379753 on Build GRCh38
rs775837288 Chr.9: 19379468-19379468 on Build GRCh38
rs759377640 Chr.9: 19379738-19379738 on Build GRCh38
rs996410052 Chr.9: 19379807-19379807 on Build GRCh38
rs1034444210 Chr.9: 19379809-19379809 on Build GRCh38
rs962076913 Chr.9: 19379845-19379845 on Build GRCh38
rs190603821 Chr.9: 19379823-19379823 on Build GRCh38
rs780432301 Chr.9: 19379545-19379545 on Build GRCh38
rs557776415 Chr.9: 19379740-19379740 on Build GRCh38
rs892869061 Chr.9: 19379479-19379479 on Build GRCh38
rs747954110 Chr.9: 19379634-19379634 on Build GRCh38
rs763941579 Chr.9: 19379644-19379645 on Build GRCh38
rs556802287 Chr.9: 19379685-19379685 on Build GRCh38
rs11541620 Chr.9: 19379574-19379574 on Build GRCh38
rs138981947 Chr.9: 19379507-19379507 on Build GRCh38
rs372950258 Chr.9: 19379472-19379472 on Build GRCh38
rs1050202904 Chr.9: 19379734-19379734 on Build GRCh38
rs150091661 Chr.9: 19379557-19379557 on Build GRCh38
rs541359300 Chr.9: 19379745-19379745 on Build GRCh38
rs779293743 Chr.9: 19379669-19379669 on Build GRCh38
rs538586826 Chr.9: 19379832-19379832 on Build GRCh38
rs781718857 Chr.9: 19379526-19379526 on Build GRCh38
rs767919541 Chr.9: 19379618-19379618 on Build GRCh38
rs753128005 Chr.9: 19379434-19379434 on Build GRCh38
rs909466295 Chr.9: 19379673-19379673 on Build GRCh38
rs887929449 Chr.9: 19379818-19379818 on Build GRCh38
rs369923035 Chr.9: 19379436-19379436 on Build GRCh38
rs764390818 Chr.9: 19379511-19379511 on Build GRCh38
rs878937098 Chr.9: 19379485-19379485 on Build GRCh38
rs746914710 Chr.9: 19379636-19379636 on Build GRCh38
rs202000854 Chr.9: 19379463-19379463 on Build GRCh38
rs757088818 Chr.9: 19379653-19379656 on Build GRCh38
rs757742319 Chr.9: 19379514-19379514 on Build GRCh38
rs766025481 Chr.9: 19379433-19379433 on Build GRCh38
rs771844493 Chr.9: 19379635-19379635 on Build GRCh38
rs374917859 Chr.9: 19379633-19379633 on Build GRCh38
rs553796427 Chr.9: 19379775-19379775 on Build GRCh38
rs751541709 Chr.9: 19379646-19379647 on Build GRCh38
rs751933360 Chr.9: 19379606-19379606 on Build GRCh38
rs1009475602 Chr.9: 19379767-19379767 on Build GRCh38
rs756183833 Chr.9: 19379622-19379622 on Build GRCh38
rs762085527 Chr.9: 19379646-19379646 on Build GRCh38
rs958821475 Chr.9: 19379811-19379811 on Build GRCh38
rs374725963 Chr.9: 19379471-19379471 on Build GRCh38
rs538652085 Chr.9: 19379588-19379588 on Build GRCh38
rs879211906 Chr.9: 19379676-19379676 on Build GRCh38
rs145798579 Chr.9: 19379623-19379623 on Build GRCh38
rs368995322 Chr.9: 19379465-19379465 on Build GRCh38
rs764971346 Chr.9: 19379447-19379448 on Build GRCh38
rs749484981 Chr.9: 19379548-19379548 on Build GRCh38
rs775148561 Chr.9: 19379643-19379643 on Build GRCh38
rs187017123 Chr.9: 19379723-19379723 on Build GRCh38
rs757760844 Chr.9: 19379529-19379529 on Build GRCh38
rs754038430 Chr.9: 19379665-19379665 on Build GRCh38
rs534964910 Chr.9: 19379656-19379656 on Build GRCh38
rs189871470 Chr.9: 19379580-19379580 on Build GRCh38
rs768997336 Chr.9: 19379696-19379696 on Build GRCh38
rs766005931 Chr.9: 19379508-19379508 on Build GRCh38
rs138427545 Chr.9: 19379578-19379578 on Build GRCh38
rs779875802 Chr.9: 19379869-19379869 on Build GRCh38
rs753949699 Chr.9: 19379627-19379627 on Build GRCh38
rs757378785 Chr.9: 19379446-19379446 on Build GRCh38
rs933016012 Chr.9: 19379862-19379862 on Build GRCh38
rs962010893 Chr.9: 19379681-19379681 on Build GRCh38
rs373378766 Chr.9: 19379443-19379443 on Build GRCh38
rs568298527 Chr.9: 19379661-19379661 on Build GRCh38
rs150265986 Chr.9: 19379435-19379435 on Build GRCh38
rs1041436552 Chr.9: 19379768-19379768 on Build GRCh38
rs376539911 Chr.9: 19379502-19379502 on Build GRCh38
rs542259344 Chr.9: 19379806-19379806 on Build GRCh38
rs991738642 Chr.9: 19379726-19379726 on Build GRCh38
rs759038953 Chr.9: 19379638-19379638 on Build GRCh38
rs200325439 Chr.9: 19379504-19379504 on Build GRCh38

View Full Product Details