Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68227701-68228201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3605040 c.1219C>T p.R407C Chr.4: 68227943-68227943 on Build GRCh38
2829014 c.1205G>A p.R402Q Chr.4: 68227957-68227957 on Build GRCh38
5912149 c.1169G>A p.G390E Chr.4: 68227993-68227993 on Build GRCh38
2829013 c.1220G>A p.R407H Chr.4: 68227942-68227942 on Build GRCh38
1430631 c.1177A>G p.K393E Chr.4: 68227985-68227985 on Build GRCh38
View More

View Full Product Details