Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105237378-105237519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
UniGene
Hs.367639
Gene Chromosome Location
Chr.4: 105145875-105279803 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Gene Symbol
TET2-AS1
Entrez Gene ID
104384744
Gene Name
TET2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.624109
Gene Chromosome Location
Chr.4: 105171354-105352881 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2952787 c.3485C>T p.S1162F Chr.4: 105237427-105237427 on Build GRCh38
6056922 c.3451G>T p.E1151* Chr.4: 105237393-105237393 on Build GRCh38
4430996 c.3472G>T p.D1158Y Chr.4: 105237414-105237414 on Build GRCh38
6494947 c.3479G>A p.G1160E Chr.4: 105237421-105237421 on Build GRCh38
1426214 c.3463T>G p.L1155V Chr.4: 105237405-105237405 on Build GRCh38
View More

View Full Product Details